Check Order Status
Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Actual product may vary
Fisherbrand
Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Borosilicate glass with silvered double walls and a convenient handle
Have Questions?
Catalog Number
Capacity (Metric)
FISFB1019611
1 L
Catalog number
FISFB1019611
Price (MYR)
3,288.00
PK
Add to cart
Capacity (Metric):
1 L
Price (MYR)
3,288.00
PK
Add to cart
Cylindrical- Wide Mouth
Borosilicate Glass
Silvered Double Walls
PE Stopper Included
For Use with Liquified Gases
Aluminum Housing
Lab Grade
Coated Metal Handle
Specifications
Color
Silver
For Use With (Application)
Liquid Nitrogen
Height (Metric)
375 mm
Includes
Handle
Capacity (Metric)
1 L
Depth (Metric) Interior
302 mm
Diameter (English) Inner
2.75 mm
Diameter (Metric) Inner
70 mm
Material
Aluminum