Reusable Glass Low-Form Griffin Beakers
Reusable Glass Low-Form Griffin Beakers
Actual product may vary
Reusable Glass Low-Form Griffin Beakers
Reusable Glass Low-Form Griffin Beakers
Fisher Catalogue
Reusable Glass Low-Form Griffin Beakers
Made of KG-33 borosilicate glass
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISFB100600600 mL
FISFB-100-5050 mL
FISFB1005050 mL
FISFB-100-100100 mL
FISFB100100100 mL
FISFB-100-150150 mL
FISFB-100-250250 mL
FISFB100250250 mL
FISFB-100-600600 mL
FISFB10010001000 mL
Catalog number FISFB100600
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
600 mL
Pour accurately and steadily with the help of a slightly flared spout. Fisherbrand™ Reusable Glass Low-Form Griffin Beakers are available in a variety of sizes and feature thick, heavy construction with a beaded top. Made of KG-33 borosilicate glass.
  • Thick, heavy construction with beaded top
  • Uniform wall thickness
  • Durable, matte-finish marking area for use with an ordinary pencil
  • Easy-to-read white graduated double scale on all sizes from 50 to 1000mL
Specifications
Capacity (English)20.28 oz.
DescriptionBeaker
Diameter (English) Outer3.46 in.
Diameter (Metric) Outer8.8 cm
Capacity (Metric)600 mL
MaterialBorosilicate Glass
For Use With (Application)Excellent Mechanical Strength and Durability, While Providing High Resistance to Chemical Attack and Thermal Shock
Graduation Range50 to 500 mL
Height (English)4.8 in.
Height (Metric)12.2 cm
IncludesSpout
Increments50 mL
Label TypeMatte Finish Marking
Quantity6 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960, ASTM E438, USP Type I
ScaleDual Scale
StyleGriffin
Unit SizePK