Check Order Status
Compartmentalized Petri Dishes
Actual product may vary
Fisherbrand
Compartmentalized Petri Dishes
Select from 2, 3, or 4-section plates with numbered sections
Have Questions?
Catalog Number
Type
FISFB087583
Tri Dish, Y-Plate
Catalog number
FISFB087583
Price (MYR)
1,113.00
CS
Add to cart
Type:
Tri Dish, Y-Plate
Price (MYR)
1,113.00
CS
Add to cart
Select from 2, 3, or 4-section plates with numbered sections
All dishes manufactured from virgin medical grade polystyrene
All dishes sterilized using e-beam sterilization
Convenient packaging in easy open sleeves
25 Sleeves/Cs.
Specifications
Closure Type
Stackable
Material
Polystyrene
Sterility
Sterile
Type
Tri Dish, Y-Plate
Edges
Raised Ridge
Packaging
24/Sleeve, 25 Sleeves/Cs.
Height (Metric)
15 mm
Diameter (Metric)
65 mm