Petri Dishes Specialty
Petri Dishes Specialty
Actual product may vary
Fisherbrand
Petri Dishes Specialty
Multiple sizes and styles for variety of applications
Have Questions?
Catalog NumberTypeSize
FISFB0875711ADish100 x 15 mm
Catalog number FISFB0875711A
Price (MYR)
1,434.00
CS
Add to cart
Type:
Dish
Size:
100 x 15 mm
Price (MYR)
1,434.00
CS
Add to cart
Multiple sizes and styles for variety of applications
  • All dishes manufactured from virgin medical grade polystyrene
  • All dishes sterilized using e-beam sterilization
  • Deep style petri dishes available - designed for increased media volumes, allowing for longer culture times
  • Contact plates available for collection in environments where contact is required to collect the sample
  • Convenient packaging in easy open sleeves
  • 25 Sleeves/Cs.
Specifications
Closure TypeSlippable
Bottom ShapeFlat
GriddedGridded
MaterialPolystyrene
ShapeSquare
SterilitySterile
Surface TreatmentCell Culture
TypeDish
Packaging20/Sleeve, 25 Sleeves/Cs.
Size100 x 15 mm
Dimensions (L x W)100 x 100 mm
Height (Metric)15 mm
Diameter (Metric)1.6 cm
Unit SizeCS