Check Order Status
Amber Wide Mouth Packer Glass Bottles with Black Phenolic Caps
Actual product may vary
Amber Wide Mouth Packer Glass Bottles with Black Phenolic Caps
Amber, USP Type III soda-lime glass ideal for storage of light sensitive samples or dry powders. Easy removal of contents
Have Questions?
Catalog Number
Capacity (Metric)
FISFB02911446A
120 mL
Catalog number
FISFB02911446A
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
120 mL
Features a wide opening, making them ideal for general purpose use
Amber color filters out ultraviolet wavelengths and can be used for light sensitive solids
Classic vitamin bottles for pharmaceutical packaging
Specifications
Capacity (Metric)
120 mL
Color
Amber
Material
Glass (Type 3)
Quantity
24
Closure Color
Black
Neck Finish
38-400
Closure Included
Yes
Closure Liner
Polyethylene Cone
Closure Material
Phenolic