Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Actual product may vary
Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Highly resistant to strong acids, organic solvents and other common laboratory reagents
Have Questions?
Catalog NumberCapacity (Metric)
FISFB012911050 mL
Catalog number FISFB0129110
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
50 mL
Rib construction permits stacking without binding
  • Special “dripless” lip
  • Base allows beaker to sit flat during use
  • Graduated in mL and oz.
Specifications
Capacity (Metric)50 mL
MaterialPolypropylene
For Use With (Application)Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
IncludesSpout
Increments5 mL
TypeBeaker