Reusable Glass Wide-Mouth Erlenmeyer Flasks
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Actual product may vary
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Fisherbrand
Reusable Glass Wide-Mouth Erlenmeyer Flasks
High resistance to heat, chemical attack, and mechanical shock
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduation RangeHeight (English)Closure Size
FISFB-501-250250 mL50 to 225 mL5.11 in.8
FISFB50110001000 mL250 to 1000 mL8.5 in.11
FISFB501125125 mL50 to 125 mL4.25 in.6
FISFB-501-500500 mL100 to 500 mL6.77 in.10
FISFB501500500 mL100 to 500 mL6.77 in.10
FISFB501250250 mL50 to 225 mL5.11 in.8
Catalog number FISFB-501-250
Price (MYR)
619.00
PK
Add to cart
Capacity (Metric):
250 mL
Graduation Range:
50 to 225 mL
Height (English):
5.11 in.
Closure Size:
8
Price (MYR)
619.00
PK
Add to cart
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks offer sturdy borosilicate glass construction with a heavy-duty, tooled-top finish. High resistance to heat, chemical attack, and mechanical shock.
  • Capacity scale measures approximate volume
  • Matte-finish ceramic marking spot takes writing and erases easily
Specifications
DescriptionTop style: Heavy duty tooled with capacity scale
Diameter (English) Outer3.03 in.
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Graduation Range50 to 225 mL
Graduations25 mL
Height (English)5.11 in.
Height (Metric)13 cm
No. of Positions1
Writing SurfaceMatte Finish Ceramic Enamel Marking
DisposableReusable
Mouth StyleWide
Diameter (Metric) Outer77 mm
Closure Size8
Unit SizePK