Sterile Polystyrene Petri Dish
Sterile Polystyrene Petri Dish
Actual product may vary
Sterile Polystyrene Petri Dish
Made from virgin polystyrene
Have Questions?
Catalog NumberDiameter (Metric)
FISAS405260 mm
Catalog number FISAS4052
Price (MYR)
1,130.00
CS
Add to cart
Diameter (Metric):
60 mm
Price (MYR)
1,130.00
CS
Add to cart
Made from virgin polystyrene, this petri dish is ideal for labs that require this type of material.
  • Made from virgin polystyrene
  • 60 x 15mm
  • Non-vented
  • Without stacking ring
  • 500/cs
Specifications
Closure TypeSlippable
MaterialPolystyrene
SterilitySterile
PackagingCase
Quantity500 Case
VentNo
Height (Metric)15 mm
Diameter (Metric)60 mm
Unit SizeCS