Easy Pierce Heat Sealing Foil
Easy Pierce Heat Sealing Foil
Actual product may vary
Easy Pierce Heat Sealing Foil
Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeDimensions (L x W)
FISAB-0559AluminumThermo-Seal85 x 135 mm
AB-0757AluminumEasy Pierce Foil85 x 135 mm
Catalog number FISAB-0559
Price (MYR)
1,703.00
PK
Add to cart
Material:
Aluminum
Product Type:
Thermo-Seal
Dimensions (L x W):
85 x 135 mm
Price (MYR)
1,703.00
PK
Add to cart
Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.
  • Suitable for PCR (requires the pressure from a screw-down heated lid)
  • Easily pierced
  • Peelable bond can be removed by hand
  • 20μm foil is even more easily pierced and is suitable for multiple layering
  • Recommended for use with DMSO
  • Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)
Specifications
ColorFoil
Solvent Or Alcohol ResistantLife Science Reagents Resistant, DMSO Resistant
MaterialAluminum
SterilityNon-sterile
AutoclavableNot Autoclavable
No. of Wells96
No. per Case100
No. per Pack100
Dimensions (L x W)85 x 135 mm
For Use With (Application)Transportation and high temperature applications such as PCR
For Use With (Equipment)ALPS 25 and ALPS 50V; AB-1443A
Product TypeThermo-Seal
Product LineAbgene
AdhesivePolypropylene
Certified RNase/DNase-freeNo
PierceableYes
Temperature Range (Metric)-80°C to +80°C, -80°C to +120°C (with Adequate Pressure)
Well ShapeAll well shapes
Plate CompatibilityPolypropylene
Incubation ConditionsGeneral
PeelableVery Strong Seal that requires stripper tool for peeling
Unit SizePK
Contents & Storage
General; Long term storage including storage at low temperatures