TOC Certified Containers
TOC Certified Containers
Actual product may vary
TOC Certified Containers
TOC Certified Containers
TOC Certified Containers
TOC Certified Containers
TOC Certified Containers
TOC Certified Containers
Fisherbrand
TOC Certified Containers
Trust our low-level certified vials for Total Organic Carbon testing and sampling. TOC-certified products are manufactured, packaged and tested to exacting standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)LevelStyleNo. per Case
FIS500572111440 mLCertified <20ppb TOCClear Glass Vial with Closure144
FIS12-100-25040 mLCertified <10ppb TOCClear Glass Vial, Certified72
Catalog number FIS5005721114
Price (MYR)
6,098.00
CS
Add to cart
Volume (Metric):
40 mL
Level:
Certified <20ppb TOC
Style:
Clear Glass Vial with Closure
No. per Case:
144
Price (MYR)
6,098.00
CS
Add to cart
Trust our low-level certified vials for Total Organic Carbon testing and sampling. Manufactured, tested and packaged by exacting standards,Thermo Scientific™ TOC Certified Containers are certified to contribute <10ppb or < 20ppb TOC as background.
Containers:
  • Available in several sizes and styles of containers, including the popular 40mL autosampler vials cleaned, certified and ready to use
  • Include Certificate of Analysis
Includes:
  • Sampling and storage containers
  • Autosampler compatible vials
  • Culture tubes
  • Low TOC water
Recommended for:
  • TOC testing
  • USP method 643 testing
  • Off-line grab sampling of high purity water
  • Storage of validation standards.
Specifications
DescriptionClear glass vial with closure, dust covers, Certified <20ppb TOC
Material (Liner)PTFE/Silicone
Diameter (English) Outer1.125 in.
ColorClear
Diameter (Metric) Outer28.58 mm
MaterialGlass
Volume (Metric)40 mL
Height (English)3.75 in.
LevelCertified <20ppb TOC
No. per Case144
StyleClear Glass Vial with Closure
Volume (English)1.35 oz.
Volume (Metric) Total40 mL
Closure TypeScrew Cap
Height (Metric)95.25 mm
Quantity144 Cs.
Certifications/ComplianceTested and certified to contribute < 20 ppb; includes Certificate of Analysis (COA)
TypeTOC Container
Closure Size24-414
Unit SizeCS