▸
Specialty Lab Glassware
▸
Distillation Labware
▸
Distillation Receiver, Dean Stark
Actual product may vary
Fisherbrand
Distillation Receiver, Dean Stark
For easy separation of water from reaction or distillation system
Have Questions?
Catalog Number
Standard Taper Joint
Capacity
Height (Metric)
Width (Metric)
FIS31-501-066
24/40
20 mL
325mm
95mm
Catalog number
FIS31-501-066
Price (MYR)
1,155.00
Each
Add to cart
Standard Taper Joint:
24/40
Capacity:
20 mL
Height (Metric):
325mm
Width (Metric):
95mm
Price (MYR)
1,155.00
Each
Add to cart
Graduated
With upper outer and lower inner standard taper joints, both of same size
PTFE stopcock with 2 mm bore
Specifications
Width (Metric)
95mm
Height (Metric)
325mm
Type
Distillation Receiver
Capacity
20 mL
Standard Taper Joint
24/40