▸
First Aid and Medical
▸
Patient Care Products
▸
Cotton Balls and Packing
▸
Nonsterile Cotton Balls
Actual product may vary
Fisherbrand
Nonsterile Cotton Balls
Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.
Have Questions?
Catalog Number
Weight (Metric)
Quantity
Size
FIS22-456-880
0.45 kg
4000 Cs.
Medium
Catalog number
FIS22-456-880
Price (MYR)
249.00
Case
Add to cart
Weight (Metric):
0.45 kg
Quantity:
4000 Cs.
Size:
Medium
Price (MYR)
249.00
Case
Add to cart
Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.
Uniform in size and shape
Made of pure, natural, bleached cotton for increased absorbency
Available in sizes medium, large, extra large and bulk roll
Specifications
Product Type
Cotton Balls
Sterility
Non-sterile
Size
Medium
Quantity
4000 Cs.
Weight (English)
0.006 lb.
Weight (Metric)
0.45 kg