Disposable Inoculating Loops and Needles
Actual product may vary
Disposable Inoculating Loops and Needles
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
Have Questions?
Catalog NumberVolumeProduct TypeQuantity
FIS22-363-6041 μLInoculating Loop and Needle, Rigid9600/Cs.
Catalog number FIS22-363-604
Price (MYR)
532.00
Pack
Volume:
1 μL
Product Type:
Inoculating Loop and Needle, Rigid
Quantity:
9600/Cs.
Price (MYR)
532.00
Pack
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and in anaerobic chambers. Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation.
  • Combi Loop™ has a 10μL loop on one end and a 1μL loop on the other
  • Convenient than metal loops; use a loop directly from the package, then throw it away
  • Gamma irradiated for sterility in accordance with U.S. Pharmacopeia Volume XX
  • No flaming required, eliminating the hazards of open flames, spattering, cross-contamination, and the risk of infection from pathogens distributed by aerosol formation
  • Color coded for easy identification
Specifications
DisposableSingle-use
Diameter (English) Inside0.035 in.
Diameter (Metric) Inside0.09 cm
Length (English)8 in.
Length (Metric)20 cm
MaterialBiologically Inert Polymer
SterilitySterile
Volume1 μL
ColorBlue
Product TypeInoculating Loop and Needle, Rigid
IncludesCertificate of dimensional accuracy
Quantity9600/Cs.
Packaging TypeZiplock Bag
Unit SizePack