Disposable Inoculating Loops and Needles, Flexible Needle and Loop
Actual product may vary
Disposable Inoculating Loops and Needles, Flexible Needle and Loop
Disposable Inoculating Loops and Needles, Flexible Needle and Loop
Disposable Inoculating Loops and Needles, Flexible Needle and Loop
Disposable Inoculating Loops and Needles, Flexible Needle and Loop
Fisherbrand
Disposable Inoculating Loops and Needles, Flexible Needle and Loop
For quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 9 in. (23cm) are molded from a specially formulated, biologically inert polymer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolumeProduct TypeColor
FIS22-363-6011 μL, 10 μLInoculating Loop, FlexibleNatural
FIS22-363-59710 μLInoculating Loop and Needle, FlexibleYellow
FIS22-363-60010 μLInoculating Loop, FlexibleYellow
Catalog number FIS22-363-601
Price (MYR)
390.00
Case
Volume:
1 μL, 10 μL
Product Type:
Inoculating Loop, Flexible
Color:
Natural
Price (MYR)
390.00
Case
For quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 9 in. (23cm) are molded from a specially formulated, biologically inert polymer.
  • Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation
  • Gamma-irradiated for sterility in accordance with U.S. Pharmacopoeia, Volume XX
  • Combi Loop has a 10μL loop on one end and a 1μL loop on the other
  • Color coded for easy identification
Specifications
DisposableSingle-use
Diameter (English) Inside0.035/0.14 in.
Diameter (Metric) Inside0.09/0.38 cm
Length (English)8.15 in.
Length (Metric)207 mm
MaterialBiologically Inert Polymer
SterilitySterile
Volume1 μL, 10 μL
ColorNatural
Product TypeInoculating Loop, Flexible
IncludesCertificate of dimensional accuracy
Quantity250/Cs.
Packaging TypePaper Peel Pouch, Individually Wrapped
Unit SizeCase