Vinyl-Coated Lead Rings
Vinyl-Coated Lead Rings
Actual product may vary
Vinyl-Coated Lead Rings
Vinyl-Coated Lead Rings
Vinyl-Coated Lead Rings
Fisherbrand
Vinyl-Coated Lead Rings
Stabilizes expensive glassware. Bel-Art™ Vinyl-Coated Lead Rings will not scratch flasks.
Have Questions?
Catalog NumberHeight (Metric)
FIS22-27830812.5 mm
Catalog number FIS22-278308
Price (MYR)
371.00
EA
Add to cart
Height (Metric):
12.5 mm
Price (MYR)
371.00
EA
Add to cart
Stabilizes expensive glassware. Bel Art™ SP Scienceware™ Vinyl-Coated Lead Rings will not scratch flasks.
  • The bright red vinyl coating is chemical-resistant and will not scratch flasks
  • Round, closed ring
Specifications
Product TypeLead Ring
AutoclavableNot Autoclavable
ColorBright Red
HoldsRound Bottom Flask (10 mL)
MaterialVinyl-coated Lead
ShapeRing
ShelvesNone
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)5 to 10mL Erlenmeyer-style straight-necked flasks
Length (Metric)51 mm
Width (Metric)51 mm
Height (Metric)12.5 mm
Diameter (English) Inner0.74 in.
Diameter (English) Outer2 in.
Diameter (Metric) Outer0.5 cm
Diameter (Metric) Inner1.89 cm
Weight (English)0.36 lb.
Weight (Metric)165 g
Unit SizeEA