Test Tube Basket
Test Tube Basket
Actual product may vary
Test Tube Basket
Polypropylene basket well-suited for rinsing glassware
Have Questions?
Catalog NumberDimensions (L x W x H)
FIS22-2605346 x 6 x 6 in.
Catalog number FIS22-260534
Price (MYR)
-
Dimensions (L x W x H):
6 x 6 x 6 in.
Polypropylene basket well-suited for rinsing glassware
  • Can also be used for transporting small laboratory animals
  • Steam autoclavable at 121°C (250°F)
Specifications
IncludesBasket and Lid
Product TypeBasket for Test Tubes
AutoclavableAutoclavable
ColorWhite
HoldsTest Tubes
MaterialPolypropylene
ShapeSquare
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)Drying Rack
Size152 x 152 x 152 mm
Dimensions (L x W x H)6 x 6 x 6 in.
Length (Metric)15.2 cm
Length (English)6 in.
Width (Metric)15 cm
Width (English)6 in.
Height (Metric)15.2 cm
Height (English)6 in.
Unit SizeEA