Spectro™ Cell Rack
Spectro™ Cell Rack
Actual product may vary
Spectro™ Cell Rack
Holds twenty 10 mm light path spectrophotometer cuvets in two tiers with numbered wells
Have Questions?
Catalog NumberHolds
FIS22-260504Light Path Cells (10 mm)
Catalog number FIS22-260504
Price (MYR)
333.00
EA
Add to cart
Holds:
Light Path Cells (10 mm)
Price (MYR)
333.00
EA
Add to cart
Holds twenty 10mm light path spectrophotometer cuvets in two tiers with numbered wells
  • The wells hold standard cuvets with a 12.5 mm cross-section in numbered positions
  • The sloped top of this polypropylene rack makes the second tier more accessible
  • Steam autoclavable at 121°C (250°F)
Specifications
Product TypeCuvet Rack
AutoclavableNot Autoclavable
ColorWhite
HoldsLight Path Cells (10 mm)
MaterialPolypropylene
Positions20
ShapeRectangular
StackableNo
For Use With (Equipment)Cuvettes
Dimensions (L x W x H)8.25 x 2.7 x 1.3 in. (20.9 x 7 x 3.5cm)
Length (Metric)20.9 cm
Length (English)8.25 in.
Width (Metric)7 cm
Width (English)2.7 in.
Height (Metric)3.5 cm
Height (English)1.3 in.
Unit SizeEA