Check Order Status
StaFreez™ Irreversible Freeze-Thaw Indicator
Actual product may vary
Fisherbrand
StaFreez™ Irreversible Freeze-Thaw Indicator
Irreversible indication of whether or not unstable frozen biological materials have been thawed
Have Questions?
Catalog Number
Temperature Range (Metric)
FIS22-246196
-20° or lower
Catalog number
FIS22-246196
Price (MYR)
836.00
PK
Add to cart
Temperature Range (Metric):
-20° or lower
Price (MYR)
836.00
PK
Add to cart
Monitors surface temperature of frozen items
Liquid-crystal indicator displays environmental temperatures items have been subjected to with color change
After activated, product is applied to frozen material at approximately -20°C
Indicator remains blue until frozen material is warmed
Upon warming, color changes first to gray, then to gray-green, and finally to black as sample temperature reaches 0°C
Color will not return to blue, even if the sample is refrozen, indicating thawing has occurred
Specifications
For Use With (Application)
Monitoring surface temperature of frozen item
Product Type
Indicator
Temperature Range (Metric)
-20° or lower