Polypropylene Graduated Pitchers
Polypropylene Graduated Pitchers
Actual product may vary
Fisher Catalogue
Polypropylene Graduated Pitchers
Constructed of translucent polypropylene, ideal for general laboratory use and offer well designed spouts and large handles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS22-163691500 mL
FIS14-513-30410 L
Catalog number FIS22-163691
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Constructed of translucent polypropylene, ideal for general laboratory use and offer well designed spouts and large handles.
  • Metric graduations molded into outside wall
  • Steam autoclavable at 121°C (250°F)
  • Balanced handle for controlled pouring
  • 5000 mL (5 L) and 10000 mL (10 L) models feature an updated molded handle for a more secure grip, and a redesigned hand-slot at base for easy lifting
Specifications
Diameter (English)Top3.2 in.
Diameter (Metric)Top8.13 cm
Capacity (Metric)500 mL
MaterialPolypropylene
For Use With (Application)For Controlled Pouring, Chemically Resistant and Shatterproof
Height (English)5.3 in.
Height (Metric)13.5 cm
Increments10 mL
Temperature (English) Max. Operating250°F
Temperature (Metric) Max. Operating121°C
TypePitcher
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
ScaleMolded Graduations
StyleGriffin, Low Form
Unit SizeEA