Cheesecloth
Cheesecloth
Actual product may vary
Fisherbrand
Cheesecloth
100% pure cotton. Excellent for wiping cuvettes and microscopes; cleaning cameras; and drying delicate glassware, syringes, pipettes, and pH electrodes.
Have Questions?
Catalog NumberDimensions (L x W)
FIS22-055053100 yd. x 36 in.
Catalog number FIS22-055053
Price (MYR)
-
Dimensions (L x W):
100 yd. x 36 in.
100% pure cotton Fisherbrand™ Cheesecloth cleans any optical grade surface.
  • Bleached
  • Grade 10 mesh size
  • Brilliant white
  • 36 in. x 100 yd.
Specifications
Product TypeCheesecloth
Length (English)100 yd.
Length (Metric)91.44 m
Width (English)36 in.
Width (Metric)91.4 cm
MaterialCotton
Certifications/ComplianceUSP
ColorWhite
Dimensions (L x W)100 yd. x 36 in.
For Use With (Application)For use to wipe cuvettes and microscopes, clean cameras, and dry delicate glassware, syringes, pipettes, and pH electrodes
Unit SizePK