OdorGone™ Autoclave Deodorizer
OdorGone™ Autoclave Deodorizer
Actual product may vary
OdorGone™ Autoclave Deodorizer
High concentration liquid deodorant in capsule form
Have Questions?
Change viewbuttonViewtableView
Catalog NumberScentQuantity
FIS22-032085Lemon25 Capsules
FIS22-032079Apple25 Capsules
Catalog number FIS22-032085
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Scent:
Lemon
Quantity:
25 Capsules
One capsule placed in each load absorbs offensive odors, leaving your autoclaves smelling fresh and clean.
Specifications
Product TypeAutoclave Capsule
Color-codedNo
FormLiquid
ScentLemon
SterilityNon-sterile
AutoclavableAutoclavable
ColorTranslucent Yellow
FormatCapsules
For Use With (Application)Autoclave Deodorizer
Quantity25 Capsules
Unit SizePK