Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring Storage Cabinets
Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring Storage Cabinets
Actual product may vary
Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring Storage Cabinets
Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring Storage Cabinets
Fisherbrand
Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring Storage Cabinets
Six-drawer model with six insert trays holds 900 embedding rings or 1800 tissue cassettes
Have Questions?
Catalog NumberProduct Type
FIS22-025591Filing Cabinet
Catalog number FIS22-025591
Price (MYR)
-
Product Type:
Filing Cabinet
Six-drawer model with six insert trays holds 900 embedding rings or 1800 tissue cassettes
  • For long-term, orderly storage, and easy retrieval of embedded cassettes or rings
  • Durable, tan metal cabinet
  • Available in six-drawer configuration for daily lab-bench usage
  • Clear plastic drawers feature removable, six-section insert trays with adjustable foam stops to keep rings or cassettes in a vertical orientation
Specifications
Product TypeFiling Cabinet
ColorClear
Holds900 Embedding Rings or 1800 Tissue Cassettes
Unit SizeEA