Search Thermo Fisher Scientific
Catalog Number | Size | Length (Metric) |
---|---|---|
FIS19-181-562 | Medium | 108 cm |
FIS19-181-560 | X-Small | 104 cm |
FIS19-181-561 | Small | 106 cm |
FIS19-181-563 | Large | 110 cm |
FIS19-181-564 | X-Large | 112 cm |
FIS19-181-565 | 2X-Large | 114 cm |