Spilfyter™ Chemical Classifiers Strip Kits
Spilfyter™ Chemical Classifiers Strip Kits
Actual product may vary
Spilfyter™ Chemical Classifiers Strip Kits
Spilfyter™ Chemical Classifiers Strip Kits
Fisherbrand
Spilfyter™ Chemical Classifiers Strip Kits
For identifying risks in unknown spilled liquids
Have Questions?
Catalog NumberTypeWeight (English)
FIS19-021915Chemical Classifier Kit1 lb.
Catalog number FIS19-021915
Price (MYR)
640.00
PK
Add to cart
Type:
Chemical Classifier Kit
Weight (English):
1 lb.
Price (MYR)
640.00
PK
Add to cart
Chemical Classifier Strips show responder if an unknown spill contains critical levels of these contaminant categories: acid/base, oxidizer, fluoride, petroleum/organic solvent, iodine/bromine/chlorine. Classifier Chart compares sample colors to standard color.
  • Recommended for all emergency response teams
  • Quickly assist responder in classifying spill for proper treatment
  • Use anywhere
  • Case or tube versions
Chemical Classifier Kit consists of:
  • Six Chemical Classifier Strips
  • One bobbin of polypropylene thread
  • Laminated Chemical Classifier Chart
  • Carrying Tube
Chemical Classifier Station consists of:
  • 20 Chemical Classifier Strips
  • Two bobbins of polypropylene thread
  • Laminated Chemical Classifier Chart
  • Carrying Case
Use strips to identify unknowns in sewers, construction sites, accident sites, storage facilities, landfills, waste management facilities.
Specifications
TypeChemical Classifier Kit
For Use With (Application)Identify unknowns in sewers, construction sites, accident sites, storage facilities, landfills, waste management facilities by using
IncludesChemical classifier strips, bobbin of polypropylene thread, laminated chemical classifier chart
Packaging Quantity1 Ea.
Storage Requirements20°C
Weight (English)1 lb.
Weight (Metric)0.4 kg
Unit SizePK