Step-On Biohazard Waste Containers
Step-On Biohazard Waste Containers
Actual product may vary
Fisherbrand
Step-On Biohazard Waste Containers
Provide safe, temporary collection of biological waste (no sharps). Foot operated pedal with self-closing lid reduces the risk of contamination. Alert workers to potential biohazards and meet OSHA requirements for exposures to bloodborne pathogens.
Have Questions?
Catalog NumberCapacity (English)Height (Metric)Length (Metric)Width (Metric)
FIS18-880A6 gal.41 cm42 cm34 cm
Catalog number FIS18-880A
Price (MYR)
980.00
EA
Add to cart
Capacity (English):
6 gal.
Height (Metric):
41 cm
Length (Metric):
42 cm
Width (Metric):
34 cm
Price (MYR)
980.00
EA
Add to cart
  • Suitable for medical facilities and research labs
  • Safely collect biological waste; reduce contamination
  • Foot lever opens container for hands-free operation
  • Heavy-wall, high density polyethylene construction won’t rust or dent
  • Labeled to advise workers of potential biohazards
Specifications
Capacity (Metric)22.7 L
Certifications/ComplianceFM, OSHA 29 CFR 1910.1030, NFPA
Diameter (Metric) Outer41 cm
Diameter (English) Outer16 in.
Capacity (English)6 gal.
ColorRed
Dimensions (L x W x H)16.5 x 13.5 x 16 in. (42 x 34 x 41 cm)
Height (Metric)41 cm
Length (Metric)42 cm
Width (Metric)34 cm
IncludesProminent biohazard warning label, foot lever
For Use With (Application)Suitable for medical facilities and research labs
Width (English)13.5 in.
Length (English)16.5 in.
Height (English)16 in.
MaterialHDPE
Product TypeBiohazard Waste Can
Unit SizeEA