EasySep™ Multistand
EasySep™ Multistand
Actual product may vary
EasySep™ Multistand
Immunomagnetic column-free magnet
Have Questions?
Catalog NumberQuantity
FIS18-0101 EasySep™ Multistand
Catalog number FIS18-010
Price (MYR)
889.00
EA
Add to cart
Quantity:
1 EasySep™ Multistand
Price (MYR)
889.00
EA
Add to cart
  • The EasySep™ Multistand allows the separation of up to 4 samples at one time
  • An ingenious reversible clamping bar allows the Multistand to hold up to 4 EasySep™ (Catalog #18000) or 4 'The Big Easy' EasySep™ Magnets (Catalog #18001)
  • After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes
  • The receiving tube bar is indexed to ensure proper position of the tubes
Specifications
For Use With (Application)Cell Isolation
Quantity1 EasySep™ Multistand
Unit SizeEA