Lab Hazard Warning Label Refills
Lab Hazard Warning Label Refills
Actual product may vary
Fisherbrand
Lab Hazard Warning Label Refills
Printed in safety colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDisplay Text
FIS15-934Flammable
FIS15-933Corrosive
Catalog number FIS15-934
Price (MYR)
16.00
EA
Add to cart
Display Text:
Flammable
Price (MYR)
16.00
EA
Add to cart
Printed in safety colors.
For use with Fisherbrand™ Lab Hazard Warning Label Dispenser (cat. no. 15931).
  • Tape rolls preprinted with warnings in safety colors: POISON (yellow on black); CORROSIVE (black on yellow); FLAMMABLE (red on white); and RADIOACTIVE (red on yellow)
  • Serrated tear-off edge for easy tape separation
  • Approximately 200 imprints per roll
Specifications
ColorAssorted
FormatTape
Product TypeFlammable Warning Refill Tape
Display TextFlammable
For Use With (Application)Printed in safety colors
Quantity4/Cs.
Length (English)57 yd.
Length (Metric)20.3 m
Width (English)0.5 in.
Unit SizeEA