Lab Hazard Warning Labels with Dispenser
Lab Hazard Warning Labels with Dispenser
Actual product may vary
Fisherbrand
Lab Hazard Warning Labels with Dispenser
Printed in safety colors.
Have Questions?
Catalog NumberColor
FIS15-925Multi-colored
Catalog number FIS15-925
Price (MYR)
319.00
EA
Add to cart
Color:
Multi-colored
Price (MYR)
319.00
EA
Add to cart
  • Dispenser supplied with four pressure-sensitive tape rolls preprinted with warnings in safety colors: POISON (yellow on black); CORROSIVE (black on yellow); FLAMMABLE (red on white); and RADIOACTIVE (red on yellow)
  • Serrated tear-off edge for easy tape separation
  • Approximately 200 imprints per roll
  • Refill rolls available separately
Specifications
ColorMulti-colored
FormatTape
Product TypeTape Dispenser
For Use With (Application)Printed in safety colors
Includes4 Rolls with preprinted warnings corrosive (black on yellow), flammable (red on white), poison (yellow on black), radioactive (red on yellow)
Quantity1 Ea.
Length (English)22 yd.
Length (Metric)20 m
Width (English)0.5 in.
Unit SizeEA