Stainless-Steel Crucible Tongs
Stainless-Steel Crucible Tongs
Actual product may vary
Fisherbrand
Stainless-Steel Crucible Tongs
Well-balanced, strong, accurately aligned
Have Questions?
Catalog NumberLength (Metric)
FIS15-18623 cm
Catalog number FIS15-186
Price (MYR)
80.00
EA
Add to cart
Length (Metric):
23 cm
Price (MYR)
80.00
EA
Add to cart
Well-balanced, strong, accurately aligned
Fisherbrand™ Stainless-Steel Crucible Tongs. Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Precision instrument joint ordinarily found only on high-quality surgical instruments
  • Carefully tapered, serrated tips meet precisely when closed
  • Withstand high temperatures
  • Won't rust
Specifications
For Use With (Application)High quality surgical instruments
IncludesPrecision closure
Length (English)9 in.
Tip StyleTapered, Serrated
Length (Metric)23 cm
MaterialStainless Steel
TypeCrucible Tongs
Unit SizeEA