Kimble™ KIMAX™ Reusable Tubes with Rubber-Lined Screw Cap
Kimble™ KIMAX™ Reusable Tubes with Rubber-Lined Screw Cap
Actual product may vary
Fisherbrand
Kimble™ KIMAX™ Reusable Tubes with Rubber-Lined Screw Cap
Black phenolic cap with inert rubber liner
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)Thread Size
FIS14-930E25 mm150 mm50 mL24-410
Catalog number FIS14-930E
Price (MYR)
1,313.00
PK
Add to cart
Diameter (Metric):
25 mm
Length (Metric):
150 mm
Capacity (Metric):
50 mL
Thread Size:
24-410
Price (MYR)
1,313.00
PK
Add to cart
Black phenolic cap with inert rubber liner
  • White marking spot
  • Cat. No. 14-930C tube is suitable for determination of insulin according to method of Alving, Rubin, and Millor (Journal of Biological Chemistry [1939], 127, 3)
Specifications
Certifications/ComplianceASTM E982, Type VI, Class A
Closure ColorBlack
Closure MaterialPhenolic
Capacity (Metric)50 mL
Length (Metric)150 mm
MaterialBorosilicate Glass
Label TypeMarking Spot
Liner MaterialRubber
Thread Size24-410
Diameter (Metric)25 mm
Closure TypeScrew Cap
Product TypeReusable Test Tube
Unit SizePK