Sterileware™ Sterile Styrene Sampler Spoons
Sterileware™ Sterile Styrene Sampler Spoons
Actual product may vary
Fisherbrand
Sterileware™ Sterile Styrene Sampler Spoons
Polystyrene plastic sterile sampler spoons are designed to gather small amounts of sample material.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FIS14-429D14.79 mL200/Cs.
FIS14-429B2.46 mL200/Cs.
FIS14-429C4.93 mL200/Cs.
Catalog number FIS14-429D
Price (MYR)
2,976.00
CS
Add to cart
Capacity (Metric):
14.79 mL
Quantity:
200/Cs.
Price (MYR)
2,976.00
CS
Add to cart
Manufactured from polystyrene plastic. SP Scienceware™ Sterileware™ Sterile Styrene Sampler Spoons are designed to gather small amounts of sample material.
  • Sterile, individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
  • FDA grade materials; safe for food, drug and cosmetic contact
  • Long handle reaches easily into jars, bottles and other confined areas, and minimizes the potential for personal contact with sampled material
  • Overall length 17.8 cm (7 in.)
Specifications
Product TypeSpoon
MaterialPolystyrene
AutoclavableNot Autoclavable
IncludesCertificate of sterilization
Handle TypeLong
Capacity (English)3 tsp.
Capacity (Metric)14.79 mL
DisposableSingle-use
SterilitySterile
ColorWhite
Certifications/ComplianceFDA
For Use With (Application)Designed to gather small amounts of sample material
Quantity200/Cs.
Dimensions (L x W)17.8 x 4 cm
Length (English)7 in.
Length (Metric)17.8 cm
Width (English)1.53 in.
Width (Metric)4 cm
Unit SizeCS