Pipette Pump™ Pipetters
Pipette Pump™ Pipetters
Actual product may vary
Fisherbrand
Pipette Pump™ Pipetters
Provides safe, accurate and trouble-free pipetting. Bel Art™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric)
FIS13-683CGreen10 mL
FIS13-683BBlue2 mL
Catalog number FIS13-683C
Price (MYR)
305.00
Each
Add to cart
Color:
Green
Volume (Metric):
10 mL
Price (MYR)
305.00
Each
Add to cart
Provides safe, accurate and trouble-free pipetting. Bel Art™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available
Specifications
Adjustable Tip SpacingNo
AutoclavableAutoclavable
ColorGreen
DisposableReusable
For Use WithPipette
Grip TypeErgonomic
MaterialPolypropylene
ModelPipette Pump Pipettors
Number of Channels1
SterilityNon-sterile
TypePipette
Volume (Metric)10 mL
For Use With (Equipment)Pipets
ResistanceAcid and Alkali
Unit SizeEach