accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
Actual product may vary
accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
Fisherbrand
accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesJunctionFilling Solution TypeLength (Metric) Cable
FIS13-620-285
also known as FIS13 -620-285
BNCSingle CeramicSP135-50076.2 cm
FSP13-620-285BNCSingle CeramicSP135-50076.2 cm
Catalog number FIS13-620-285
also known as FIS13 -620-285
Price (MYR)
2,054.00
Each
Connector Types:
BNC
Junction:
Single Ceramic
Filling Solution Type:
SP135-500
Length (Metric) Cable:
76.2 cm
Price (MYR)
2,054.00
Each
For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical
  • 0 to 14pH range
  • Porous ceramic junction
  • Twist-open/twist-close cap for easy filling
  • Constant distance between FS-5 Universal Glass pH bulb and reference junction means high reproducibility
  • Immersion depth, ∽4 in. (10.1cm)
  • 30 in. long (76.2cm) leads, except 13-620-91, which has 60 in. long (152.4cm) lead
Choice of Ag/AgCl Single- or Double-Junction internal reference elements
  • Single-junction models with Ag/AgCl reference are perfect for routine pH and titration measurements of aqueous solutions with small samples, small vessels
  • Double-junction models with Ag/AgCl reference are useful in aqueous and nonaqueous solutions, and in applications requiring TRIS buffers
  • For aqueous and nonaqueous samples
  • Double-junction Ag/AgCl eliminate measurement and clogging problems when used in research with Tris buffers
Specifications
Closure TypeTwist Open/Twist Close
ColorBlue
Connector TypesBNC
JunctionSingle Ceramic
MaterialGlass
pH Range0 to 14
Temperature (Metric) Operating23°C to 212°C
TypeCombination pH Electrode
Filling Solution TypeSP135-500
For Use With (Application)For routine pH and titration measurements with small samples and small vessels, where separate half cell electrodes may be impractical
For Use With (Equipment)pH meter
IncludesRemovable bulb protector
Internal ReferenceAg/AgCl
Response Time<10 sec.
Temperature (English) Operating-5°F to +100°F
Temperature (Metric) Range-5°C to +100°C
Warranty1 Year
Length (English)4 in.
Length (English) Cable30 in.
Length (Metric)101.6 mm
Length (Metric) Cable76.2 cm
Unit SizeEach