Glass Kinematic Viscometer Tubes
Glass Kinematic Viscometer Tubes
Actual product may vary
Fisherbrand
Glass Kinematic Viscometer Tubes
Cannon™-Fenske type viscometer tubes for measuring kinematic viscosity of petroleum products by ASTM™ D 445.
Have Questions?
Catalog NumberApproximate ConstantASTM NumbersSizeDescription
FIS13-616G0.5 cSt/sec.350350Uncalibrated
Catalog number FIS13-616G
Price (MYR)
1,179.00
EA
Add to cart
Approximate Constant:
0.5 cSt/sec.
ASTM Numbers:
350
Size:
350
Description:
Uncalibrated
Price (MYR)
1,179.00
EA
Add to cart
Cannon™-Fenske type viscometer tubes for measuring kinematic viscosity of petroleum products by ASTM™ D 445.
  • Available calibrated (with calibration certificate) or uncalibrated
  • Excellent precision
  • Borosilicate glass
  • Specifications also conform with ASTM D 446
  • Minimum sample volume, 7mL
  • Requires liquid bath with depth of 203mm
Specifications
Product TypeCannon-Fenske Routine Viscometer
Viscosity Range100 to 500 cSt
CalibratedNo
DescriptionUncalibrated
Approximate Constant0.5 cSt/sec.
ASTM Numbers350
IncludesInstruction sheet
MaterialBorosilicate Glass
Depth (English) Bath8 in.
Depth (Metric) Bath20.3 cm
AccuracyASTM D445, ASTM D446, ISO 3104, ISO 3105
Precision±0.2%
For Use With (Application)For measuring kinematic viscosity of transparent Newtonian liquids, particularly petroleum products or lubricants
Size350
Volume (Metric)7 mL
Unit SizeEA