Stainless-Steel Utility Trays
Stainless-Steel Utility Trays
Actual product may vary
Fisherbrand
Stainless-Steel Utility Trays
Corrosion resistant and easy to clean
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)Type
FIS13-361D20.75 x 12.75 x 2.5 in.Catheter Tray
FIS13-361C16.5 x 10 x 2.5 in.Instrument Tray
FIS13-361B12.25 x 7.75 x 2.25 in.Instrument Tray
Catalog number FIS13-361D
Price (MYR)
-
Dimensions (L x W x H):
20.75 x 12.75 x 2.5 in.
Type:
Catheter Tray
Corrosion resistant and easy to clean
Specifications
Capacity (Metric)7.5 L
Dimensions (L x W x H)20.75 x 12.75 x 2.5 in.
Height (Metric)6.4 cm
Length (Metric)53 cm
MaterialStainless Steel
TypeCatheter Tray
Width (Metric)32 cm
Length (English)20.75 in.
Height (English)2.5 in.
ShapeRectangular
Width (English)12.75 in.
Capacity (English)7.125 qt.
Unit SizeEA