CoorsTek™ Clear Glass Mortar and Pestle Sets
CoorsTek™ Clear Glass Mortar and Pestle Sets
Actual product may vary
Fisherbrand
CoorsTek™ Clear Glass Mortar and Pestle Sets
Provide clear view of contents
Have Questions?
Catalog NumberCapacityDiameter (English) InnerHeight (English)
FIS12-970B4 oz., 118.29mL3.93 in.3.66 in.
Catalog number FIS12-970B
Price (MYR)
188.00
EA
Add to cart
Capacity:
4 oz., 118.29mL
Diameter (English) Inner:
3.93 in.
Height (English):
3.66 in.
Price (MYR)
188.00
EA
Add to cart
  • Mortar has heavy molded base and lip
  • With clear glass pestle
  • Offer clear view of materials
Specifications
Capacity (Metric)118.29 mL
Capacity (English)4 oz.
Diameter (English) Inner3.93 in.
Diameter (English) Outer3.9 in.
Diameter (Metric) Inner10cm
Diameter (Metric) Outer9.9cm
Product TypeMortar and Pestle
MaterialFlint Glass
Height (English)3.66 in.
Height (Metric)9.3cm
Capacity4 oz., 118.29mL
Unit SizeEA