Cover Glasses: Squares, Various Thicknesses
Cover Glasses: Squares, Various Thicknesses
Actual product may vary
Cover Glasses: Squares, Various Thicknesses
Made of finest optical borosilicate glass, with uniform thickness and size
Have Questions?
Catalog NumberThickness (Metric)Length (Metric)Width (Metric)
FIS12-540B#2 (0.17 to 0.25 mm)22 mm22 mm
Catalog number FIS12-540B
Price (MYR)
-
Thickness (Metric):
#2 (0.17 to 0.25 mm)
Length (Metric):
22 mm
Width (Metric):
22 mm
Made of finest optical borosilicate glass, with uniform thickness and size
Square shape.
  • Corrosion-resistant
  • Lint-free and ready to use
  • Available in squares, circles and rectangles
  • Sold by weight; comes in a pack of 1 oz.
Specifications
Product TypeCoverslip
ShapeSquare
MaterialBorosilicate Glass
Length (Metric)22 mm
Width (Metric)22 mm
Thickness (Metric)#2 (0.17 to 0.25 mm)
For Use With (Application)Microscope, histology, cytology, hematology, microbiology
Quantity1 oz./Pk.
Unit SizePK