Baume Hydrometers
Baume Hydrometers
Actual product may vary
Fisherbrand
Baume Hydrometers
Baume heavy scale for working with liquids heavier than water.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/ComplianceIncrementsLength (Metric)Baume Range
FIS11-571EWithout 3-point certification0.1°290 mm40 to 51°
FIS11-571BWithout 3-point certification0.1°305 mm9 to 21°
Catalog number FIS11-571E
Price (MYR)
159.00
EA
Add to cart
Certifications/Compliance:
Without 3-point certification
Increments:
0.1°
Length (Metric):
290 mm
Baume Range:
40 to 51°
Price (MYR)
159.00
EA
Add to cart
Baume heavy scale for working with liquids heavier than water.
  • Permanently marked with individual serial numbers
Specifications
TypeBaume Hydrometer
Increments0.1°
Certifications/ComplianceWithout 3-point certification
Length (Metric)290 mm
MaterialSoda Lime Glass
Baume Range40 to 51°
For Use With (Application)Baume 'Heavy' Scale For Liquids Heavier than Water
Unit SizeEA