Precision Specific Gravity 150 - 185mm Long Hydrometers
Precision Specific Gravity 150 - 185mm Long Hydrometers
Actual product may vary
Precision Specific Gravity 150 - 185mm Long Hydrometers
Requires 140mL liquid volume for submersion.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/ComplianceLength (Metric)Specific Gravity Range
FIS11-556PNIST, ASTM E100, ASTM E126170 mm1.480 to 1.550
FIS11-556HNIST, ASTM E100, ASTM E126170 mm1.120 to 1.190
Catalog number FIS11-556P
Price (MYR)
155.00
EA
Add to cart
Certifications/Compliance:
NIST, ASTM E100, ASTM E126
Length (Metric):
170 mm
Specific Gravity Range:
1.480 to 1.550
Price (MYR)
155.00
EA
Add to cart
Requires 140mL liquid volume for submersion.
Used where a high degree of accuracy is required in hydrometer readings
  • Permanently marked with individual serial numbers
  • Accuracy meets or exceeds ASTM E 100 specifications
  • Calibrated according to ASTM E 126 specifications
  • With divisions of 0.001
  • Use Cylinder 08-530E, available separately
Specifications
Increments0.001
Certifications/ComplianceNIST, ASTM E100, ASTM E126
Length (Metric)170 mm
MaterialSoda Lime Glass
For Use With (Application)Measure the True Specific Gravity of Liquids that are Lighter than Water
Specific Gravity Range1.480 to 1.550
Volume (Metric)140 mL
Unit SizeEA