Specific Gravity Hydrometers
Specific Gravity Hydrometers
Actual product may vary
Fisherbrand
Specific Gravity Hydrometers
Measure true specific gravity of liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/ComplianceIncrementsLength (Metric)Specific Gravity Range
FIS11-520FASTM E100, ASTM E1260.005290 mm1.000 to 1.600
FIS11-510BASTM E100, ASTM E1260.001310 mm0.800 to 0.910
FIS11-510CASTM E100, ASTM E1260.001305 mm0.900 to 1.000
FIS11-510DASTM E100, ASTM E1260.005305 mm0.650 to 1.000
FIS11-512CASTM E100, ASTM E1260.002180 mm0.900 to 1.000
FIS11-520AASTM E100, ASTM E1260.002305 mm1.000 to 1.220
FIS11-520BASTM E100, ASTM E1260.002275 mm1.200 to 1.420
FIS11-520CASTM E100, ASTM E1260.002285 mm1.400 to 1.620
Catalog number FIS11-520F
Price (MYR)
155.00
EA
Add to cart
Certifications/Compliance:
ASTM E100, ASTM E126
Increments:
0.005
Length (Metric):
290 mm
Specific Gravity Range:
1.000 to 1.600
Price (MYR)
155.00
EA
Add to cart
Measure true specific gravity of liquids.
  • Chemical-resistant bulbs and stems
  • Streamlined to avoid trapping air bubbles
  • With directional device that stops the instrument with the scale facing the user for more convenient, more accurate readings
  • Double scales can be read without turning the hydrometer.
Specifications
TypeSpecific Gravity Hydrometer
Increments0.005
Certifications/ComplianceASTM E100, ASTM E126
Length (Metric)290 mm
For Use With (Equipment)300mL Borosilicate Glass cylinder
MaterialSoda Lime Glass
For Use With (Application)Measure true specific gravity of liquids heavier than Water
Specific Gravity Range1.000 to 1.600
Volume (Metric)250 mL
Unit SizeEA