Check Order Status
Perforated Glass Beads
Actual product may vary
Fisherbrand
Perforated Glass Beads
Spread boiling area
Have Questions?
Catalog Number
Diameter (Metric)
FIS11-311C
6 mm
Catalog number
FIS11-311C
Price (MYR)
238.00
PK
Add to cart
Diameter (Metric):
6 mm
Price (MYR)
238.00
PK
Add to cart
Soda-lime glass
Packed in 16 oz. (454g) bottles
Specifications
Material
Soda Lime Glass
Diameter (Metric)
6 mm
Quantity
454 g