Imhoff Settling Cone
Actual product may vary
Imhoff Settling Cone
For measuring sand and other settled matter in water and sewage.
Have Questions?
Catalog NumberVolume (Metric)
FIS10-987-0541000 mL
Catalog number FIS10-987-054
Price (MYR)
509.00
Each
Volume (Metric):
1000 mL
Price (MYR)
509.00
Each
  • Molded from glass-clear, unbreakable styrene acrylonitrile
  • With continuous graduations up to 1000mL
  • For tests involving high volume of settled matter
  • Leakproof polyethylene plug permits easy withdrawal of sediment for gravimetric analysis and simplifies cleaning
  • Cones graduated in divisions of: 0 to 2mL in 0.1mL, 2 to 10mL in 0.5mL, 10 to 40mL in 1mL, 40 to 100mL in 2mL, and 100 to 1000mL in 50mL
Specifications
DescriptionFor tests involving high volume of settled matter
Volume (Metric)1000 mL
For Use With (Application)For measuring sand and other settled matter in water and sewage
GraduatedYes
IncludesLeakproof polyethylene plug
MaterialGlass-clear, unbreakable styrene acrylonitrile
TypeImhoff settling cone
Dimensions (L x W)Dia. x H: 12 x 50 cm (4.7 x 19.7 in.)
Unit SizeEach