PYREX™ Buchner Funnels with Fine Porosity Fritted Disc
PYREX™ Buchner Funnels with Fine Porosity Fritted Disc
Actual product may vary
PYREX™ Buchner Funnels with Fine Porosity Fritted Disc
Useful where filter paper would be attacked by material being filtered
Have Questions?
Catalog NumberCapacity (Metric)PorosityPore Size
FIS10-358E30 mLMedium10 to 15 μm
Catalog number FIS10-358E
Price (MYR)
864.00
EA
Add to cart
Capacity (Metric):
30 mL
Porosity:
Medium
Pore Size:
10 to 15 μm
Price (MYR)
864.00
EA
Add to cart
Useful where filter paper would be attacked by material being filtered
These funnels are especially useful where filter paper would be attacked by the material being filtered. The disc should not be subjected to pressure differential in excess of 1 atmosphere. The top edge is beaded.
Specifications
Capacity (Metric)30 mL
Diameter (Metric) Stem8 mm
MaterialBorosilicate Glass
Height (Metric)139 mm
Certifications/ComplianceASTM E128
For Use With (Application)Useful where filter paper would be attacked by material being filtered
Diameter (Metric) Disc30 mm
IncludesFritted discs
Length (Metric) Stem75 mm
PorosityMedium
Pore Size10 to 15 μm
Product TypeBuchner Funnel
Unit SizeEA