HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ T, 60-80 mesh
HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ T, 60-80 mesh
Actual product may vary
Fisherbrand
HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ T, 60-80 mesh
HayeSep™ T was used together with a thermoconductivity detector to study the solubility of trioxane in methanol at different temperatures and to measure the vapour pressure data for this binary mixture.
Have Questions?
Catalog number FIS10-309
Price (MYR)
520.00
PK
Add to cart
Price (MYR)
520.00
PK
Add to cart
HayeSep™ porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.
HayeSep is a registered trademark of Hayes Separation Inc.
Specifications
Particle Size60/80 Mesh
PackagingBottle
Quantity75 cc
Surface Area (Metric)˜250 m2/g
Product LineHayeSep
For Use With (Application)GC
Density˜0.38 g/mL (free fall density)
Unit SizePK