HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ N, 80-100 mesh
HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ N, 80-100 mesh
Actual product may vary
HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ N, 80-100 mesh
HayeSep™ N was used as a adsorbent for analytical preconcentration of methane which contributes majorly to the greenhouse effect. It was found to have good adsorption capacity, highest overall selectivity and highest enthalpy for methane adsorption as compared to other adsorbents.
Have Questions?
Catalog number FIS10-295
Price (MYR)
14.00
EA
Add to cart
Price (MYR)
14.00
EA
Add to cart
Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. HayeSep™ has a high surface area owing to its micropores thereby making it a suitable candidate for separation of gases and volatile organic compounds (VOCs). Furthermore they are relatively inert and exhibit hydrophobicity. HayeSep™ N is a copolymer of poly(divinylbenzene-co-ethylenedimethacrylate).
HayeSep is a registered trademark of Hayes Separation Inc.
Specifications
Particle Size80/100 Mesh
PackagingBottle
Quantity75 cc
Surface Area (Metric)405 m2/g
Product LineHayeSep
For Use With (Application)GC
Density˜0.36 g/mL (free fall density)
Unit SizeEA