Kimble™ KIMAX™ Brand Class A Volumetric Flasks
Kimble™ KIMAX™ Brand Class A Volumetric Flasks
Actual product may vary
Kimble™ KIMAX™ Brand Class A Volumetric Flasks
Calibrated To Contain and To Deliver
Have Questions?
Catalog number FIS10-218B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Two graduation lines
  • 50mL comes with standard taper 9 stopper; 100mL, standard taper 13
Specifications
CalibrationCalibrated To Contain, To Deliver
Closure MaterialGlass
Capacity (Metric)100 mL
MaterialBorosilicate Glass
Height (English)7.08 in.
Height (Metric)18 cm
Tolerance±0.08 mL
GraduatedYes
Closure Size13
Unit SizeCS