Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper
Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper
Actual product may vary
Fisherbrand
Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper
Single graduation ring blasted on the neck
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FIS10-205F1000 mL22
FIS10-205B50 mL9
FIS10-205C100 mL13
FIS10-205D250 mL16
FIS10-205E500 mL19
Catalog number FIS10-205F
Price (MYR)
785.00
EA
Add to cart
Capacity (Metric):
1000 mL
Closure Size:
22
Price (MYR)
785.00
EA
Add to cart
Single graduation ring blasted on the neck
  • Calibrated “To Contain”
  • Calibration indefinite if not exposed to hydrofluoric acid, hot phosphoric acid, or strong, hot alkalis
  • Do not heated above 150°C when dry
Specifications
CalibrationCalibrated To Contain
Closure MaterialGlass
Capacity (Metric)1000 mL
MaterialGlass
No. per Case1
Tolerance±0.3 mL
GraduatedYes
Closure Size22
Unit SizeEA