▸
Flasks
▸
Volumetric Flasks
▸
Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper
Actual product may vary
Fisherbrand
Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper
Single graduation ring blasted on the neck
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Closure Size
FIS10-205C
100 mL
13
FIS10-205B
50 mL
9
FIS10-205D
250 mL
16
FIS10-205E
500 mL
19
FIS10-205F
1000 mL
22
5 Options
Catalog number
FIS10-205C
Price (MYR)
259.00
Each
Add to cart
Capacity (Metric):
100 mL
Closure Size:
13
Price (MYR)
259.00
Each
Add to cart
Single graduation ring blasted on the neck
Calibrated “To Contain”
Calibration indefinite if not exposed to hydrofluoric acid, hot phosphoric acid, or strong, hot alkalis
Do not heated above 150°C when dry
Specifications
Calibration
Calibrated To Contain
Closure Material
Glass
Capacity (Metric)
100 mL
Material
Glass
No. per Case
1
Tolerance
±0.08 mL
Graduated
Yes
Closure Size
13