Hygienic Hi-Flex Wall Bracket System
Hygienic Hi-Flex Wall Bracket System
Actual product may vary
Fisherbrand
Hygienic Hi-Flex Wall Bracket System
This hygienic, modular wall bracket provides a flexible solution for storing your cleaning tools and utensils in any area of your site. The rail easily attaches to four wall mounts by sliding it on from the right or left side.
Have Questions?
Catalog NumberColor
FIS10-115White
Catalog number FIS10-115
Price (MYR)
728.00
PK
Add to cart
Color:
White
Price (MYR)
728.00
PK
Add to cart
  • Grip band modules and hook modules can be clicked on the rail and locked in place using one of many small nothces in the rail
  • Up to ten hanging modules can be placed on the rail (or up to five grip band modules)
  • System comes with three grip band modules and two hooks
  • Easy to dissemble for cleaning and replacement
Specifications
Product TypeWall Bracket
ColorWhite
MaterialPolypropylene
Height (English)1.9 in.
Length (English)16.5 in.
Weight (English)0.63 lb.
Width (English)3.4 in.
Unit SizePK