Kimble™ KIMAX™ Culture Media Flasks
Kimble™ KIMAX™ Culture Media Flasks
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Culture Media Flasks
Screw-thread finish
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-093AA50 mL
Catalog number FIS10-093AA
Price (MYR)
-
Capacity (Metric):
50 mL
Screw-thread finish
  • Includes long caps with cemented-in white rubber liners
Specifications
Diameter (English) Outer2 in.
Capacity (Metric)50 mL
MaterialBorosilicate Glass
For Use With (Application)Used to mix and store culture media, as well as with other culturing techniques
Graduation Range20 to 50 mL
Graduations10 mL
Height (English)3 in.
Height (Metric)7.8 cm
IncludesCap and white Rubber liner
Thread Size24-410
Closure TypeScrew
Diameter (Metric) Outer5 cm
Unit SizePK